Would you like to log in again?
UBC13 Peptide, Rockland Immunochemicals
Supplier: Rockland Immunochemical
000-001-342
RL000-001-342EA 142.94 USD InStock
RL000-001-342
UBC13 Peptide, Rockland Immunochemicals
Proteins and Peptides
UBC13 Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
UBC13 peptide for use in control assays with purified antibodies Anti-UBC13 (p/n 600-101-342).
Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty