Would you like to log in again?
Huntington pS421 Control Phospho Peptide, Rockland Immunochemicals
Supplier: Rockland Immunochemical
000-000-433
RL000-000-433
Huntington pS421 Control Phospho Peptide, Rockland Immunochemicals
Huntington pS421 Phospho Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Huntingon pS421 phospho peptide for use in control assays with purified antibodies Anti-Huntingon pS421 (p/n 600-401-433). Also available Huntingon non-phospho S421 peptide (p/n 000-000-450).
Intended for use as a control in peptide competition assays to demonstrate the binding properties of anti-Huntingon pS421 antibody to bind Huntington protein in the presence or absence of control peptide. Control peptide should be used at 1.0 µg per 1.0 µg of antibody per assay.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty