Would you like to log in again?
Latex Gloves, Powder Free, American Cleanstat Durham
Supplier: THOMAS SCIENTIFIC TE
XC1002-L XC1002-XL
76238-810CS 226.96 USD InStock
76238-810 76238-812
Latex Gloves, Powder Free, American Cleanstat Durham
Gloves
The XtraClean series powder free latex gloves are specifically designed for use in controlled environments.
  • Ambidextrous
  • Silicone free
  • Beaded Cuff
  • Natural Color
The XC1002 line is cleanroom processed and packaged for use in Class 100 cleanrooms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty