Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pink Anti-Static Bags, Elkay Plastics

Supplier: Elkay Plastics
FAS62436 FAS61014 FAS61016 FAS61824
This is choice when static control is necessary but Faraday cage protection is not required.

  • Surface resistivity of 1012 ohms/sq.in.
  • Bags can be heat-sealed or taped and are amine-free

Effectively eliminate the hazard of static damage to sensitive electronic components during packaging, storing and shipping.



 

ORDER

Expand All / Collapse All
Dimensions Thickness Weight Supplier No. VWR Catalog Number Unit Price Quantity
10×14" 6.0 mil 56.0 lbs./m FAS61014 76245-894 Case of 500 Retrieving
10×16" 6.0 mil 64.3 lbs/m FAS61016 76244-248 Case of 500 Retrieving
18×24" 6.0 mil 173.6 lbs/m FAS61824 76244-250 Case of 250 Retrieving
24×36" 6.0 mil 347.3 lbs./m FAS62436 76244-252 Case of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty