Would you like to log in again?
Amersham Imager 680 Blot and Gel Imagers, GE Healthcare
Supplier: GE Healthcare - Life Sciences
29270772 29270770 29270771 29270769
103568-420EA 59630.77 USD InStock
103568-420 103568-416 103568-418 103568-414
Amersham Imager 680 Blot and Gel Imagers, GE Healthcare
Photodocumentation Systems
A sensitive, robust, and easy-to-use chemiluminescence CCD imager designed to provide publication-quality digital images of DNA and protein gels and blots.
  • High sensitivity
  • Sharp images with high resolution
  • Easy to use
  • Highly flexible performance
  • Robust design
Detect low abundant protein molecules easily. Advanced settings allow up to 50 incremental exposures and exposure times up to 10 h. Choose from four binning modes, including “no binning” for sharper images and improved resolution of closely spaced bands—great for chemiluminescence imaging!
Intuitive software that’s easy to learn and use with on-board analysis functions. Adjust sample position and focus when needed.
Image a chemiluminescent sample and a colored molecular weight marker simultaneously with automatic overlay. The overlay image retains the marker color for easy identification of molecular weights.
Choose from five imaging options to cover all of your imaging needs from colorimetric to chemiluminescence to fluorescence.
Low maintenance design provides dependability for high-throughput, multi-user labs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty