Would you like to log in again?
Nano Soldering Handle, JBC Tools USA
Supplier: JBC TOOLS USA INC. TE
NT105-A
76243-534EA 190.82 USD InStock
76243-534
Nano Soldering Handle, JBC Tools USA
Soldering Supplies Soldering Irons
The smallest handle for soldering very small-sized components requiring the greatest precision.
  • Shortest distance Tip-to-Grip
  • Soft grip
Provides greater manual control of the tool, including when using a microscope. The grip offer a soft and comfortable feel and prevent the fingers from sliding down.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty