Would you like to log in again?
LaserTab Series Tamper Labels, Brady
Supplier: Brady Worldwide
29761
76241-596PK 1403.15 USD InStock
76241-596
LaserTab Series Tamper Labels, Brady
Labels
The tamper evident labels provide outstanding performance in laser and xerographic printing processes.
  • Chemical and corrosion resistant
  • Smear and weather resistant
  • Provides permanency and smudge resistance with dot-matrix printing
Ideal for use in areas where evidence of tampering is required. Not recommended for barcode printing.
Rectangle in shape, Laser toner receptive, Tamper evident are the material properties.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty