Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol 200 Proof ACS, USP, Ph. Eur., BP, JP, FCC

Danger
Synonyms: Ethyl alcohol

111WORLD200PL05M 111WORLD200PL05 111WORLD200CS4L 111WORLD200DM55M 111WORLD200DM55UV
76289-722EA 614.52 USD InStock
76289-722 76289-720 76289-718 76289-682 76289-724
Ethanol 200 Proof ACS, USP, Ph. Eur., BP, JP, FCC
Ethanol


Formula: C₂H₅OH
MW: 46,07 g/mol
Boiling Pt: 78.3 °C (1013 hPa)
Melting Pt: -117 °C
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: ambient
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty