Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water USP, Ph. Eur., JP

Supplier: Greenfield

Synonyms: Dihydrogen oxideIce

232WORLDDM55UV
76289-648EA 521.15 USD InStock
76289-648
Water USP, Ph. Eur., JP
Water


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-778-1
UN: 0000
Merck Index: 13,10098
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty