Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-1,2-Propanediol Kosher USP, Ph. Eur., JP, FCC

Supplier: Greenfield

Synonyms: Propylene glycol(±)-1,2-Propylene glycol(±)-1,2-Dihydroxy propane

369WORLDCS4L 369WORLDDM55M 369WORLDPL05
76289-708CS 808.1 USD InStock
76289-708 76289-712 76289-710
(±)-1,2-Propanediol Kosher USP, Ph. Eur., JP, FCC
Propylene glycol


Formula: CH₃CH(OH)CH₂OH
MW: 76,1 g/mol
Boiling Pt: 187.6 °C (1013 hPa)
Melting Pt: -59 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://us.vwr.com/stibo/web/std.lang.all/05/57/710557.jpg
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty