Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥99% ACS, USP, FCC, Ph. Eur., BP, JP

Supplier: Greenfield

Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-Propanol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

231WORLDPL05M 231WORLDCS4L 231WORLDDM55M 231WORLDPL05 231WORLDDM55UV
76289-630EA 532.5 USD InStock
76289-630 76289-626 76289-634 76289-628 76289-632
2-Propanol ≥99% ACS, USP, FCC, Ph. Eur., BP, JP
2-Propanol


Formula: (CH₃)₂CHOH
MW: 60,1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://us.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty