Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ACS, USP, NF, BP, Ph. Eur.

Supplier: Greenfield

Danger
Synonyms: Ethyl ethanoateAcetidinVinegar naphthaEthyl acetic esterAcetic acid ethyl esterAcetic etherAcetoxyethane

330WORLDCS4L 330WORLDDM55M 330WORLDPL05M 330WORLDPL05
76289-600CS 564.4 USD InStock
76289-600 76289-728 76289-726 76289-602
Ethyl acetate ACS, USP, NF, BP, Ph. Eur.
Ethyl acetate


Formula: CH₃COOC₂H₅
MW: 88,11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://us.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty