Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol 191 Proof ACS, USP, Ph. Eur., BP, JP

Danger
Synonyms: Ethyl alcohol

111WORLD191CS4L 111WORLD191DM55M 111WORLD191PL05M 111WORLD191DM55UV 111WORLD191PL05
76289-694CS 682.81 USD InStock
76289-694 76289-702 76289-698 76289-700 76289-696
Ethanol 191 Proof ACS, USP, Ph. Eur., BP, JP
Ethanol


Formula: C₂H₅OH
MW: 46,07 g/mol
Boiling Pt: 78.3 °C (1013 hPa)
Melting Pt: -117 °C
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: ambient
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty