Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Propanol ACS, FCC, Ph. Eur.

Supplier: Greenfield

Danger
Synonyms: Propanol-1EthylcarbinolPropyl alcohol1-Propyl alcoholn-Propyl alcoholEthyl carbinolPropan-1-ol1-Hydroxy propanen-Propanoln-Propan-1-ol1-HydroxypropanePropylic alcoholn-Propyl Alcohol

334WORLDDM55M
76289-646EA 2105.1 USD InStock
76289-646
1-Propanol ACS, FCC, Ph. Eur.
1-Propanol


Formula: H₃CCH₂CH₂OH
MW: 60,1 g/mol
Boiling Pt: 97 °C (1013 hPa)
Melting Pt: -127 °C
Density: 0.8053 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
Merck Index: 13,07934
https://us.vwr.com/stibo/web/std.lang.all/16/15/731615.jpg
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty