Human Recombinant BCA-1
Supplier: MBL International
JM-4001-1000 JM-4001-100
101736-036EA 944.78 USD
101736-036 101736-712
Human Recombinant BCA-1
Proteins and Peptides
Human BCA-1 (B Cell-Attracting chemokine 1), also known as BLC or CXCL13, is a recently identified new CXC chemokines. Human BCA-1 is a highly effective attractant for human blood B lymphocytes, but was inactive on freshly isolated or IL-2 stimulated T lymphocytes, monocytes and neutrophils.
  • The human BCA-1 is a 10.1 kDa protein containing 85 amino acid residues
Alternative names: B cell attracting chemokine-1, CXCL13, BLC (mouse), BLR1 ligand, Angie, B lymphocyte chemoattractant, CXC chemokine BLC, small-inducible cytokine B13.