Z-VDVAD-FMK, MBL International

Supplier: MBL INTERNATIONAL CORP MS

JM-1142-1 JM-1142-5
101974-710EA 403.08 USD
101974-710 101974-712
Z-VDVAD-FMK, MBL International
Proteins and Peptides
A synthetic peptide that irreversibly inhibits activity of caspase-2. The inhibitor is designed as a methyl ester to facilitate cell permeability. The product can be used for both in vitro and in vivo studies.

Research area: Apoptosis

Shelf life: One year under proper storage conditions

Caution: For research purpose only! Not to be used in humans. If the intended use is on purified or recombinant enzymes, esterase should be added to generate free carboxyl groups.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR