Recombinant Caspase-1 Inhibitor

Supplier: MBL International
BV-1012-3
101974-178EA 254.06 USD
101974-178
Recombinant Caspase-1 Inhibitor
Proteins and Peptides
YVAD is the preferred recognition sequence for ICE/caspase-1. The inhibitor is designed as a methyl ester to facilitate cell permeability.

Sequence: Z-Tyr-Val-Ala-Asp(OMe)-FMK (FMK, fluoromethyl ketone)

Concentration: 2 mM

Caution: For research use only. Not for use in diagnostic purposes.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR