Recombinant Caspase-6 Inhibitor

Supplier: MBL International
BV-1011-3
101974-174EA 241.75 USD
101974-174
Recombinant Caspase-6 Inhibitor
Proteins and Peptides
VEID is the specific recognition sequence for caspase-6/Mch2. The inhibitor is designed as a methyl ester to facilitate cell permeability.

Sequence: Z-Val-Glu(OMe)-Ile-Asp(OMe)-FMK (FMK, fluoromethyl ketone)

Concentration: 2 mM

Caution: For research use only. Not for use in diagnostic purposes.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR