Desco’s Stat-A-Rest® Full Coverage Foot Grounders, Menda
4596
76246-730PR 86.89 USD
76246-730
Desco’s Stat-A-Rest® Full Coverage Foot Grounders, Menda
Desco’s Stat-A-Rest® full coverage foot grounders provides most coverage of any foot grounder.
Full coverage foot grounders have a walking voltage generation of 50 Volts (Conductive Flooring) and 100 Volts (with Dissipative Flooring).
Certifications: Tested per ANSI/ESD STM97.2.