Would you like to log in again?
LaCrosse Wireless Rain Station
Supplier: LA CROSSE TECHNOLOGY
724-1710V2
470309-886
LaCrosse Wireless Rain Station
Discover how we impact the quality of the air.
  • Self-emptying Rain Bucket-Empties Via Internal Tilting Cups
  • Rainfall History Graph: View Last 7 Days, Weeks, or Months on Graph
  • Monitor Indoor and Wireless Outdoor Temperature
  • Rainfall Alert and Precipitation Alarm
This LaCrosse Wireless Rain Gauge features a self-emptying rain collector that wirelessly transmits rainfall and outdoor temperature data to the indoor display. The console displays temperature in either Fahrenheit or Celsius formats, the time with the day of the week and a graph for rainfall amounts. Know how much water you are using to water your lawn, or find out what the latest storm brought to your home.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty