Would you like to log in again?
EasyView™ Hygro-Thermometer and Datalogger
Supplier: FLIR COMMERCIAL SYSTEMS INC SE
EA25
470310-232EA 403.65 USD InStock
470310-232
EasyView™ Hygro-Thermometer and Datalogger
Data Loggers
The Hygro-Thermometer and Datalogger datalogs up to 15000 records for PC download.
  • Large LCD displays any two parameters simultaneously
  • Data Hold, Relative, Min/Max, Programmable power off timer
  • Stores up to 99 manual readings for display recall or up to 15000 readings for PC download
  • Programmable Start/Stop datalogging time and sample rate of 1 to 86000 seconds
Display any two parameters (Temperature and Humidity or Dew Point, or Wet Bulb) simultaneously. Min/Max function stores and recalls highest and lowest readings and displays the elapsed time to the minimum and maximum readings. Programmable auto power off. Switchable °C/°F temperature units. Data Hold function freezes reading on display.
Delivery information: Supplied complete with built-in stand, protective holster, and probe with 39" coiled cable, software and optically isolated RS-232 cable, and 6 AAA batteries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty