Would you like to log in again?
rMouse IL-5 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010001V42-0020 010001V42-0100 010001V42-0005
75929-330 75929-328 75929-332
rMouse IL-5 Protein, Rockland Immunochemicals
Interleukin-5 (IL-5) is a hematopoietic growth factor expressed in Th2, mast cells and eosinophils. IL-5 acts through its receptor, the IL-5 receptor (IL-5R) and is involved in B cell growth and eosinophil activation. IL-5 has been shown to be and is regulated by GATA-3, in addition to other transcription factors. Human and mouse IL-5 are cross-reactive. Recombinant mouse IL-5 is a non-glycosylated, disulfide-linked homodimer, containing two 113 amino acid chains, with a total molecular weight of 26.2 kDa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty