Would you like to log in again?
KDM5B protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010-001-S36 010-001-S36S
75929-428EA 665.18 USD InStock
75929-428 75929-426
KDM5B protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
KDM5B is a lysine (K)-specific demethylase 5B which is highly expressed in testis and shows the lowest expression in skeletal muscle (1). KDM5B is a histone H3 lysine 4 (H3K4) demethylase up-regulated in prostate cancer and associated with androgen receptor where it regulated its transcriptional activity in reporter gene assays (2). Overexpression of KDM5B in HeLa cells results in loss of tri-, di-, and monomethyl H3K4, but does not alter the methylation status of H3K9 or H3K36. A KDM5B containing highly conserved putative DNA/chromatin binding motif is specifically up-regulated in breast cancer. KDM5B Protein is ideal for investigators involved in Signaling Proteins, Deacetylase/Demethylase Proteins, Cancer, Cell Cycle, and Inflammation research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty