Would you like to log in again?
RANTES Mouse Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010-001-C20 010-001-C20S
RL010001C20 RL010001C20S
RANTES Mouse Recombinant Protein, Rockland Immunochemicals
Rantes, in the chemokine CC family, is a secreted chemoattractant for blood monocytes, memory T-helper cells and eosinophils. Causes the release of histamine from basophils and activates eosinophils. It is very T-cell and macrophage specific. Recombinant Mouse Rantes produced in E.coli is a single, non-glycosylated polypeptide chain containing 68 amino acids and having a molecular mass of 7876 Daltons.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty