Would you like to log in again?
IL-6 Mouse Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010-001-310
RL010-001-310EA 311.76 USD InStock
RL010-001-310
IL-6 Mouse Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
Interleukin (IL)-6, also know as BCDG, BCGF and BSF-2, is an important proinflammatory and immunoregulatory cytokine expressed by various cells. Interleukin-6 has been shown to inhibit the growth of early stage and to promote the proliferation of advanced stage melanoma cells in vitro. Recombinant Mouse IL-6 produced in E. coli is a single, non-glycosylated polypeptide chain containing 187 amino acids and having a molecular mass of 21,709 Daltons.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty