Human Recombinant CCL13 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-V77-0005
75930-066EA 139.92 USD
75930-066
Human Recombinant CCL13 (from E. coli)
Proteins and Peptides
Monocyte Chemotactic Protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as IL-1 and TNFα. MCP-4 is a ligand for three different G protein coupled receptors, CCR2, CCR3 and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils and basophils and this signaling is associated with the allergic response. Recombinant human MCP-4 is a non-glycosylated protein, containing 74 amino acids, with a molecular weight of 8.5 kDa.