Would you like to log in again?
rHuman CCL8 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001V75-0002 009001V75-1000
75930-046EA 135.05 USD InStock
75930-046 75930-036
rHuman CCL8 Protein, Rockland Immunochemicals
Proteins and Peptides
Monocyte Chemotactic Protein 2 (MCP-2), also known as CCL8, is implicated in allergic responses through its ability to activate mast cells, eosinophils, and basophils. MCP-2 is known to signal through several G protein-coupled receptors including, CCR1, CCR2B, and CCR5. Recombinant human MCP-2 is a non-glycosylated protein, containing 76 amino acids, with a molecular weight of 8.9 kDa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty