Human Recombinant GDF5 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-U94-1000
10801-576EA 2178.59 USD
10801-576
Human Recombinant GDF5 (from E. coli)
Proteins and Peptides
Growth/Differentiation Factor 5 (GDF-5) is a growth factor that regulates cell proliferation and differentiation in embryonic and adult tissues. GDF-5 is part of the TGF family of proteins and is closely related to the BMP family of proteins. Recombinant human GDF-5 is a non-glycosylated homodimer, containing two 120 amino acids chains, with a total molecular weight of 27.4 kDa. To enable bacterial expression the N-terminal sequence of Ala-Pro-Leu-Thr was replaced with a Lys.