Would you like to log in again?
N6AMT2 protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S54S 009-001-S54
75930-200EA 338.75 USD InStock
75930-200 75930-202
N6AMT2 protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
N6AMT2 is a putative DNA methyltransferase, belongs to the methyltransferase superfamily, AML1 family. The N6AMT2 gene is conserved in human, mouse, rat, chimpanzee, dog, cow, chicken, zebrafish, fruit fly, mosquito, C.elegans, S.cerevisiae, K.lactis, N.crassa, A.thaliana, and rice (1). The N6AMT2 phosphorylation at Ser-2 has been reported (2). The function of this protein has not been widely studied yet. N6AMT2 Protein is ideal to investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Apoptosis/Autophagy, Cancer, Cell Cycle, and Neurobiology research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty