Would you like to log in again?
MGMT protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S47S 009-001-S47
75929-936EA 353.41 USD InStock
75929-936 75929-938
MGMT protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
MGMT or O-6-methylguanine-DNA methyltransferase is the major mutagenic and carcinogenic lesion in DNA induced by simple alkylating mutagens. The methylation of the MGMT promoter in gliomas is a useful predictor of the responsiveness of the tumors to alkylating agents (1). The clinical trial substantiates the predictive value of O-6-methylguanine-DNA methyltransferase promoter methylation in glioblastoma patients treated with temozolomide (2). Methylation of the MGMT promoter in tumors is associated with longer survival in glioblastoma patients. MGMT Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Apoptosis/Autophagy, Cancer, Cell Cycle, and Neurobiology research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty