Would you like to log in again?
LOXL3 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S42 009-001-S42S
75929-672 75929-670
LOXL3 protein-GST fusion, Rockland Immunochemicals
LOXL3 is a member of the lysyl oxidase gene family. This family is essential to the biogenesis of connective tissue. The lysyl oxidase gene family encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin (1). LOXL3 is highly expressed in central nervous system, uterus, heart, and leukocyte (1). LOXL3 plays an important role in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis. LOXL3 Protein is ideal for investigators involved in Signaling Proteins, Lysyl Oxidase Proteins, Cancer, and Metabolic Disorder research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty