Would you like to log in again?
LOXL2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S41 009-001-S41S
75929-668 75929-666
LOXL2 protein-GST fusion, Rockland Immunochemicals
LOXL2 is a member of the lysyl oxidase gene family which is essential for the biogenesis of connective tissue. LOXL2 encodes an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. LOXL2 may play an important role in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family (1). LOXL2 may mediate fibroblast activation through enzymatic crosslinking of fibrillar collagen and local matrix tention, resulting in activation of TGFb1 signaling (2). LOXL2 Protein is ideal for investigators involved in Signaling Proteins, Lysyl Oxidase Proteins, Cancer, and Metabolic Disorder research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty