Would you like to log in again?
KDM4A protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S33 009-001-S35 009-001-S35S
75929-416EA 665.18 USD InStock
75929-416 75929-424 75929-422
KDM4A protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
KDM4A or lysine (K)-specific demethylase 4A is a member of the Jumonji domain 2 (JMJD2) family that encodes a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. KDM4A functions in euchromatin to remove histone methylation marks that are associated with active transcription (1). KDM4A also act as a trimethylation-specific demethylase those converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor (2). KDM4A Protein is ideal for investigators involved in Cancer, Cell Cycle, and Inflammation research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty