Would you like to log in again?
IRAK1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S22 009-001-S22S
75929-222EA 665.18 USD InStock
75929-222 75929-220
IRAK1 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
IRAK1 encodes the interleukin-1 receptor-associated kinase 1, one of two putative serine/threonine kinases that become associated with the interleukin-1 receptor (IL1R) upon stimulation. The exposure of HeLa cells or human embryonic kidney cells overexpressing IL1R to IL1 caused rapid association of IRAK with the IL1R complex and phosphorylation of IRAK (1).IRAK1is partially responsible for IL1-induced upregulation of the transcription factor NF-kappa B. IRAK1 as a risk gene with critical role in the pathogenesis of systemic lupus erythematosus (2). IRAK1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Inflammation, NfkB Pathway, p38 Pathway, and Ser/Thr Kinases research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty