Would you like to log in again?
FRS3 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R96 009-001-R96S
10801-506 10801-504
FRS3 protein-GST fusion, Rockland Immunochemicals
FRS3 or fibroblast growth factor receptor substrate 3 is a peripheral plasma membrane protein that is a substrate for the fibroblast growth factor receptor. During FGF or NGF stimulation, FRS3 becomes tyrosine phosphorylated and then serves as a platform for the recruitment of multiple signaling proteins for activation of the Ras-MAP kinase signaling cascade (1). FRS3 is mainly expressed in fibroblast and myoblast cell lines and undergo robust tyrosine phosphorylation in response to several mitogenic ligands. Through interaction with Rho family of small GTPases, FRS3 has been implicated in the reorganization of the actin cytoskeleton and subsequent morphological changes in various cells (2). FRS3 Protein is ideal for investigators involved in Signaling Reagents, Protein Substrates, AKT/PKB Pathway, Angiogenesis, Cancer, and ERK/MAPK Pathway research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty