Human Recombinant FOS (from Baculovirus (Sf9 Insect cells))
Supplier: Rockland Immunochemicals
009-001-R95S 009-001-R95
10802-946EA 713.69 USD
10802-946 10802-944
Human Recombinant FOS (from Baculovirus (Sf9 Insect cells))
Proteins and Peptides
FOS is a member of the FOS gene family that consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. FOS gene encodes a leucine zipper protein that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation (1). In some cases, expression of the FOS gene has also been associated with apoptotic cell death. FOS overexpression leads to decreased phosphorylation and dimerization of STAT1, which in turn downregulates p21 gene expression (2). This regulatory pathway may enhance the proliferation of lymphocytes in rheumatoid arthritis patients. FOS Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, ERK/MAPK Pathway, Invasion/Metastasis, and Neurobiology research.