Would you like to log in again?
FKBP1A protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R94S 009-001-R94
10801-424EA 353.41 USD InStock
10801-424 10801-426
FKBP1A protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
FKBP1A is a member of the immunophilin protein family which plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP1A is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and Rapamycin. FKBP1A interacts with various type I receptors, including the TGF-beta type I receptor and competitive binding assays show that the type I receptor may be a natural ligand for FKBP1A (1). FKBP1A-deficient mice have normal skeletal muscle but show severe dilated cardiomyopathy and ventricular septal defects that mimick a human congenital heart disorder (2). FKBP1A Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Cardiovascular Disease, Inflammation, Neurobiology, and NfkB Pathway research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty