Would you like to log in again?
EIF2S1 protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R86 009-001-R86S
10802-588 10802-586
EIF2S1 protein-HIS Epitope, Rockland Immunochemicals
EIF2S1, also known as eukaryotic translation initiation factor 2, subunit 1 alpha, is a 35kDa protein in which The translation initiation factor EIF2 catalyzes the first regulated step of protein synthesis initiation and promotes the binding of the initiator tRNA to 40S ribosomal subunits. Binding occurs as a ternary complex of methionyl-tRNA, EIF2, and GTP (1). The Ser51 in the mature human EIF2-alpha protein acts as the sole site of phosphorylation that leads to repression of protein synthesis. EIF2-alpha also plays an essential role in erythropoiesis (2). EIF2S1 Protein is ideal for investigators involved in Signaling Reagents, Protein Substrates, Angiogenesis, Cellular Stress, Inflammation, Neurobiology, and Ser/Thr Kinases research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty