Would you like to log in again?
DLK protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R84S 009-001-R84
10801-148EA 353.41 USD InStock
10801-148 10801-150
DLK protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
DLK or mitogen-activated protein kinase kinase kinase 12 is a transmembrane protein containing six epidermal growth factor repeats and is a serine/threonine protein kinase. DLK is involved in the differentiation of several cell types, including adipocytes. DLK plays a role of a tumor suppressor protein and is predominately expressed in neuronal cells. DLK as an activator of the Jnk pathway. Overexpression of DLK results in activation of Jnk1 and the accumulation of a hyperphosphorylated form of c-Jun (1). Postnatal loss of DLK in stem cells and niche astrocytes has been shown to regulate neurogenesis (2). DLK Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, ERK/MAPK Pathway, Invasion/Metastasis, Neurobiology, and Ser/Thr Kinases research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty