Human Recombinant CCNY (from Baculovirus (Sf9 Insect cells))
Supplier: Rockland Immunochemicals
009-001-R83 009-001-R83S
10801-074EA 713.69 USD
10801-074 10801-072
Human Recombinant CCNY (from Baculovirus (Sf9 Insect cells))
Proteins and Peptides
Cyclin Y belongs to the highly conserved cyclin family. The members of the cyclin family are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclin Y plays an important role in the processes of transcription, DNA repair, and cell cycle progression. Cyclin Y regulates cyclin-dependent kinases (1). Cyclin Y is an isoform of CCNY that is also known as CFP1 variant A which has a calculated molecular mass of 36.9 kD. (2). Cyclin Y specifically activates transcription through the MYC promoter. Cyclin Y Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.