Would you like to log in again?
CyclinO protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R82 009-001-R82S
10801-070EA 665.18 USD InStock
10801-070 10801-068
CyclinO protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
Cyclin O belongs to the highly conserved cyclin family. These family members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclin O plays an important role in the processes of transcription. Cyclin O also plays an important role in DNA repair and cell cycle progression. Cyclin O localizes exclusively in the nucleus (1). Cyclin O shows significant uracil-DNA glycosylase activity in which the activity of in vitro translated UDG2 was not as robust as that of the mitochondrial isoform of UNG (2). Cyclin O Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty