Would you like to log in again?
CyclinE1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R79 009-001-R79S
10800-970 10801-064
CyclinE1 protein-GST fusion, Rockland Immunochemicals
Cyclin E1 belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle that functions as regulators of CDK kinases. Cyclin E1 activity is required for centrosome duplication during S phase and this mechanism could coordinate centrosome reproduction with cycles of DNA synthesis and mitosis (1). The downregulation of cyclin E-CDK2 kinase activity following the G1/S-phase transition that is necessary for the maintenance of karyotypic stability. Cyclin E also has a modular centrosomal-targeting domain which is essential for promoting S phase entry in a Cdk2-independent manner (2). Cyclin E1 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty