Would you like to log in again?
CyclinC protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R77 009-001-R77S
10801-058 10801-056
CyclinC protein-GST fusion, Rockland Immunochemicals
Cyclin C is a member of the cyclin family of proteins and interacts with the cyclin-dependent kinase 8 (CDK8) and induces the phophorylation of the carboxy-terminal domain of the large subunit of RNA polymerase II. The mRNA levels of Cyclin C fluctuate during the cell cycle and peak at the G1 phase. Cyclin C acts as a critical regulator of the G0/G1 transition of human Hematopoietic stem cells (1). Cyclin C can also combine with CDK3 to stimulate Rb phosphorylation at S807/811 during the G0/G1 transition, and this phosphorylation is required for cells to exit G0 efficiently (2). CyclinC Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty