Would you like to log in again?
CyclinA2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R76S 009-001-R76
10801-052EA 353.41 USD InStock
10801-052 10801-054
CyclinA2 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
CyclinA2 belongs to the highly conserved cyclin family whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle and function as regulators of CDKs. CyclinA2 is an essential component of all embryonic and somatic cell cycles in mammals (1). CyclinA2 is highly expressed in the testis and certain myeloid leukemia cells (2). Cyclin A also plays a major role in the control of DNA replication (3). CyclinA2 binds and activates CDC2 or CDK2 and thus promotes both cell cycle G1/S and G2/M transitions. CyclinA2 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty