Human Recombinant CDK5R1 (from Baculovirus (Sf9 Insect Cells))
Supplier: Rockland Immunochemicals
009-001-R64 009-001-R64S
10800-722EA 364.21 USD
10800-722 10800-724
Human Recombinant CDK5R1 (from Baculovirus (Sf9 Insect Cells))
Proteins and Peptides
CDK5P25 (cyclin-dependent kinase 5, regulatory subunit 1) is a neuron-specific activator of CDK5 and is required for proper development of the central nervous system (1). The p35 form of this protein is proteolytically cleaved by calpain, generating a p25 form of the protein which relocates from the cell periphery to nuclear and perinuclear regions. CDK5P25 regulates CDK5 activity by prolonging its activation and changing its cellular location and this may be important in pain signaling (2). The CDK5P25 accumulates in the brain neurons of patients with Alzheimer's disease and this accumulation correlates with an increase in CDK5 kinase activity which may result in aberrantly phosphorylated forms of the microtubule-associated protein tau. CDK5P25 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.