Would you like to log in again?
CDK2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R63 009-001-R63S
10800-720EA 665.18 USD InStock
10800-720 10802-302
CDK2 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
CDK2 is a member of the Cyclin-Dependent Kinase family that is ubiquitously expressed. CDK2 is a catalytic subunit of the cyclin-dependent protein kinase complex, whose activity is restricted to the G1-S phase, and essential for cell cycle G1/S phase transition. CDK2 associates with and is regulated by the regulatory subunits of the complex including Cyclin A or E, CDK inhibitor p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B) (1). CDK2 phosphorylates multiple cellular substrates including SMAD3 and FOXO1. Phosphorylation of FOXO1 leads to its inhibition (2). CDK2 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty