Would you like to log in again?
CBP protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R59S 009-001-R59
10800-620EA 353.41 USD InStock
10800-620 10800-622
CBP protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
CBP or CREB-binding protein is a nuclear transcriptional coactivator protein that binds specifically to the PKA-phosphorylated form of the CREB protein. Microinjection of an anti-CBP antiserum into fibroblasts leads to inhibition of transcription from a cAMP promoter (1). CBP can also cooperates with upstream activators, such as JUN. When JUN is phosphorylated at the transcriptionally stimulatory sites ser73 and ser63, it binds CBP with comparable affinity to CREB. Insulin signaling may directly regulate many cAMP signaling pathways at the transcriptional level by controlling CBP recruitment (2). Mutant CBP can be aberrantly recruited to CREB protein, resulting in inappropriate activation of gluconeogenesis and glucose intolerance. CBP Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cardiovascular Disease, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, Metabolic Disorder, Neurobiology, NfkB Pathway, and PKA/PKC Pathway research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty