Human Recombinant BUB1 (from Baculovirus (Sf9 Insect cells))
Supplier: Rockland Immunochemicals
009-001-R54S 009-001-R54
10800-492EA 364.21 USD
10800-492 10800-494
Human Recombinant BUB1 (from Baculovirus (Sf9 Insect cells))
Proteins and Peptides
BUB1 is a kinase which involved in spindle checkpoint function that in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint. BUB1 is highly expreesed in thymus, ovary, and testis (1). BUB1 maintains the steady-state level and centromeric localization of SGO1, and they concluded that BUB1 protects centromeric cohesion in mitosis through SGO1 (2). BUB1-mediated phosphorylation of CDC20 is required for proper checkpoint signaling and that inhibition of APC by BUB1 may partly account for the sensitivity of the spindle checkpoint. BUB1 kinase creates a mark for shugoshin localization and the correct partitioning of chromosomes. BUB1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.