Human Recombinant BCL2 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R49 009-001-R49S
10800-302EA 367.51 USD
10800-302 10800-304
Human Recombinant BCL2 (from E. coli)
Proteins and Peptides
BCL2 gene encodes an integral inner mitochondrial membrane protein that acts as an antiapoptotic protein (1). The protein BAD can antagonize both the cell cycle and antiapoptotic functions of BCL2 through binding to the BH3 domain. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma (2). BCL2 is phosphorylated on specific serine/threonine residues within the unstructured loop in response to diverse stimuli and such phosphorylation has been associated with the loss of the biological function of BCL2. BCL2 Protein is ideal for investigators involved in Singaling Proteins, Apoptosis Proteins, Apoptosis/Autophagy, Cancer, Cellular Stress, and Neurobiology research.