Would you like to log in again?
BAD protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R45 009-001-R45S
10801-966EA 665.18 USD InStock
10801-966 10801-964
BAD protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
BAD is a member of the BCL-2 family of proteins that are known to be regulators of programmed cell death. BAD is a pro-apoptotic protein that forms a heterodimer complex with BCL-xL and BCL-2 which reverses the prosurvival activity of these proteins (1). The proapoptotic activity of BAD is regulated through its phosphorylation and this inhibits the pro-apoptosis function of BAD. Protein kinases such as AKT, RAF and RSK1 can phosphorylate BAD and RSK1-induced phosphorylation of BAD at ser112 suppresses BAD-mediated apoptosis in neurons. BAD inhibits G(1) to S phase transition in MCF7 breast cancer cells and overexpression of BAD inhibits cell growth as well as cyclin D1 expression (2). BAD Protein is ideal for investigators involved in Signaling Proteins, Apoptosis Proteins, Apoptosis/Autophagy, Cancer, Cellular Stress, and Neurobiology research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty